Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB35 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543792
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543792 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543792 Supplier Invitrogen™ Supplier No. PA543792
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RTITSTYYRG THGVIVVYDV TSAESFVNVK RWLHEINQNC DDVCRILVGN Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

RAB35 possesses GTPase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB35
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RAB35
Gene Accession No. Q15286
Gene Alias 9530019H02Rik; AU040256; GTP-binding protein RAY; H-ray; I79_001225; RAB1C; Rab35; RAB35, member RAS oncogene family; Ras-related protein Rab-1C; ras-related protein rab-1c (GTP-binding protein ray); Ras-related protein Rab-35; RAY
Gene Symbols RAB35
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human RAB35.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11021
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.