Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB35 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | RAB35 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
RAB35 Polyclonal specifically detects RAB35 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
RAB35 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
11021 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
GTP-binding protein RAY, H-ray, RAB1C, RAB35, member RAS oncogene family, Ras-related protein Rab-1C, ras-related protein rab-1c (GTP-binding protein ray), ras-related protein Rab-35, RAY | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-201 of human RAB35 (NP_006852.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title