Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB35 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987995
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7995 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7995 Supplier Bioss Supplier No. BS7765R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RAB35 possesses GTPase activity.

Specifications

Antigen RAB35
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RAB35
Gene Accession No. Q15286
Gene Alias 9530019H02Rik; AU040256; BOS_16565; GTP-binding protein RAY; H-ray; I79_001225; LOC100354066; RAB1C; RAB35; RAB35, member RAS oncogene family; RAB35, member RAS oncogene family-like; Ras-related protein Rab-1C; ras-related protein rab-1c (GTP-binding protein ray); Ras-related protein Rab-35; RAY
Gene Symbols RAB35
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human RAB35.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11021
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.