Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB36 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104075
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104075 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104075 Supplier Invitrogen™ Supplier No. PA5104075
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RAB36.

Protein transport. Probably involved in vesicular traffic. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB36
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RAB36
Gene Accession No. O95755, Q8CAM5
Gene Alias 6330593L16Rik; AW047545; Rab36; RAB36, member RAS oncogene family; ras-related protein Rab-36; small GTP-binding protein Rab36
Gene Symbols RAB36
Host Species Rabbit
Immunogen A synthesized peptide derived from human RAB36(Accession O95755), corresponding to amino acid residues V35-A85.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 76877, 9609
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.