Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB36 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA537207
Description
This antibody detects endogenous protein at a molecular weight of 37 kDa. Purity is >95% by SDS-PAGE.
Protein transport. Probably involved in vesicular traffic. [UniProt].
Specifications
| RAB36 | |
| Polyclonal | |
| Unconjugated | |
| RAB36 | |
| 6330593L16Rik; AW047545; Rab36; RAB36, member RAS oncogene family; ras-related protein Rab-36; small GTP-binding protein Rab36 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 690407, 76877, 9609 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95755, Q8CAM5 | |
| RAB36 | |
| A synthetic peptide corresponding to residues in human RAB36. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction