Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB37 Antibody (OTI2F1), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273772IR
Description
RAB37 Monoclonal specifically detects RAB37 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
RAB37 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
RAB37 | |
Full length recombinant protein of human RAB37 (NP_783865) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI2F1 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
FLJ32507, member of RAS oncogene family, RAB37, member RAS oncogene family, ras-related protein Rab-37 | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
326624.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction