Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB37 Monoclonal Antibody (OTI2D1)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA526110
Description
RAB37 Monoclonal Antibody for Western Blot, ICC/IF
Rab proteins are low molecular mass GTPases that are critical regulators of vesicle trafficking. For additional background information on Rab proteins.
Specifications
RAB37 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96AX2 | |
RAB37 | |
Full length human recombinant protein of RAB37 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Western Blot, Immunocytochemistry | |
OTI2D1 | |
Unconjugated | |
RAB37 | |
B230331O03Rik; B230354I04Rik; GTPase Rab37; LOC100364984; Rab37; RAB37, member of RAS oncogene family; RAB37, member RAS oncogene family; ras-related protein Rab-37; rCG35079-like | |
Mouse | |
Affinity Chromatography | |
RUO | |
326624 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction