Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB37 Monoclonal Antibody (OTI2D1), TrueMAB™, OriGene
SDP

Catalog No. 502539764
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-253-9764 30 μL
50-167-5834 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-253-9764 Supplier OriGene Supplier No. TA505340S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RAB37 Monoclonal antibody specifically detects RAB37 in Human samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen RAB37
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI2D1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene RAB37
Gene Accession No. Q96AX2
Gene Alias B230331O03Rik; B230354I04Rik; GTPase Rab37; LOC100364984; Rab37; RAB37, member of RAS oncogene family; RAB37, member RAS oncogene family; ras-related protein Rab-37; rCG35079-like
Host Species Mouse
Immunogen Full length human recombinant protein of human RAB37 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 326624
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.