Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB38 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106718
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106718 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106718 Supplier Invitrogen™ Supplier No. PA5106718
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RAB38.

May be involved in melanosomal transport and docking. Involved in the proper sorting of TYRP1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB38
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Rab38
Gene Accession No. P57729, Q8QZZ8
Gene Alias 2310011F14Rik; AU043391; chocolate; cht; melanoma antigen NY-MEL-1; NY-MEL-1; R; RAB38; Rab38, member of RAS oncogene family; RAB38, member RAS oncogene family; rab38a; RAB38a, member RAS oncogene family; Rab-related GTP-binding protein; ras-related protein Rab-38; red eyed dilution; rrGTPbp; Ruby; ruby or red eyed dilution; small GTP binding protein
Gene Symbols Rab38
Host Species Rabbit
Immunogen A synthesized peptide derived from human RAB38(Accession P57729), corresponding to amino acid residues Y79-C129.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23682, 252916, 72433
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.