Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB38 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501727819 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-7819 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-7819 Supplier Proteintech Group Inc Supplier No. 122341AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May be involved in melanosomal transport and docking. Involved in the proper sorting of TYRP1.

Specifications

Antigen RAB38
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene RAB38
Gene Accession No. P57729, Q8QZZ8
Gene Alias Melanoma antigen NY MEL 1, NY MEL 1, RAB38, Ras related protein Rab 38, rrGTPbp
Gene Symbols Rab38
Host Species Rabbit
Immunogen RAB38 Fusion Protein Ag2874
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23682, 72433
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.