Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB39 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299624JF549
Description
RAB39 Polyclonal specifically detects RAB39 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
RAB39 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
rab-39, RAB39, member RAS oncogene family, RAB39A, rab-related GTP-binding protein, ras-related protein Rab-39A | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human RAB39. | |
0.1 mL | |
Signal Transduction | |
54734 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction