Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB39A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560004
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560004 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560004 Supplier Invitrogen™ Supplier No. PA560004
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SQRQVTREEA EKLSADCGMK YIETSA Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 92%.

The Ras-related superfamily of guanine nucleotide binding proteins includes the R-Ras, Rap, Ral/Rec and Rho/Rab subfamilies, all of which are thought to play an important role in either endocytosis or in biosynthetic protein transport. The process of transporting newly synthesized proteins from the endoplasmic reticulum (ER) to various stacks of the Golgi complex and to secretory vesicles involves the movement of carrier vesicles and requires Rab protein function. Rab proteins are also an integral part of endocytic pathways. Rab 39, also known as Rab 39A, is a 217 amino acid protein that belongs to the small GTPase superfamily. Rab 39 may be involved in vesicular trafficking.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB39A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RAB39A
Gene Accession No. Q14964
Gene Alias C230094F14Rik; Rab39; rab-39; RAB39, member RAS oncogene family; Rab39a; RAB39A, member RAS oncogene family; rab-related GTP-binding protein; ras-related protein Rab-39A
Gene Symbols RAB39A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RAB39A. Recombinant protein control fragment (Product #RP-98931).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54734
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.