Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB39B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531147
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531147 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531147 Supplier Invitrogen™ Supplier No. PA531147
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: PC-3, U87-MG. Predicted reactivity: Mouse (99%), Rabbit (100%), Rhesus Monkey (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the Rab family of proteins. Rab proteins are small GTPases that are involved in vesicular trafficking.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB39B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.86 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene RAB39B
Gene Accession No. Q8BHC1, Q96DA2
Gene Alias 6330580M05Rik; MRX72; Rab39b; RAB39B, member RAS oncogene family; ras-related protein Rab-39B; ras-related protein Rab-39B; LOW QUALITY PROTEIN: ras-related protein Rab-39B; WSMN
Gene Symbols RAB39B
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RAB39B. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116442, 67790
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.