Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB39B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543994
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543994 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543994 Supplier Invitrogen™ Supplier No. PA543994
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MEAIWLYQFR LIVIGDSTVG KSCLIRRFTE GRFAQVSDPT VGVDFFSRLV Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 92%; Zebrafish: 100%.

This gene encodes a member of the Rab family of proteins. Rab proteins are small GTPases that are involved in vesicular trafficking.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB39B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RAB39B
Gene Accession No. Q96DA2
Gene Alias 6330580M05Rik; MRX72; Rab39b; RAB39B, member RAS oncogene family; ras-related protein Rab-39B; ras-related protein Rab-39B; LOW QUALITY PROTEIN: ras-related protein Rab-39B; WSMN
Gene Symbols RAB39B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human RAB39B (aa 1-50).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116442
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.