Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rab3A Antibody (OTI1G3), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273773UV
Description
Rab3A Monoclonal specifically detects Rab3A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Rab3A | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
RAB3A | |
Full length recombinant protein of human Rab3A (NP_002857) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1G3 | |
Western Blot, Immunohistochemistry | |
RAB3A, member RAS oncogene family, RAS-associated protein RAB3A, ras-related protein Rab-3A | |
Mouse | |
Affinity Purified | |
Membrane Trafficking and Chaperones, Signal Transduction | |
5864.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction