Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB3A Mouse anti-Human, Clone: 5B7, Bioss
SDP

Catalog No. 501979843
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-197-9843 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-197-9843 Supplier Bioss Supplier No. BSM51311M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Rab proteins belong to the Ras small GTPase superfamily. The GTP-binding Rab3 subfamily has 4 known isoforms: 3A, 3B, 3C, and 3D. These proteins are expressed in highest concentrations in brain and endocrine tissues and involved in regulated exocytosis of hormones and neurotransmitters. Specifically, Rab 3s are thought to play a key role in presynaptic vesicle trafficking and priming.

Specifications

Antigen RAB3A
Applications Flow Cytometry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 5B7
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 40% glycerol and 0.05% sodium azide; pH
Gene RAB3A
Gene Accession No. P20336
Gene Alias C18A3.6; RAB3; RAB3A; RAB3A, member RAS oncogene family; RAS-associated protein RAB3A; Ras-related protein Rab-3A; ras-related protein Rab-3A-like protein; Ras-related small GTP binding protein 3A
Gene Symbols RAB3A
Host Species Mouse
Immunogen This RAB3A antibody is generated from a mouse immunized with a recombinant protein.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5864
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.