Learn More
Invitrogen™ RAB3A Recombinant Rabbit Monoclonal Antibody (PSH01-08)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA549607
Description
Sequence Similarities: 100% Mouse/Rat. Tissue Specificity: Specifically expressed in brain. Positive Control: Human brain tissue lysate, mouse brain tissue lysate, rat brain tissue lysate, human cerebellum tissue, mouse cerebellum tissue, rat cerebellum tissue, SH-SY5Y, N2A, PC-12. Subcellular Location: Cytoplasm, cytosol, Lysosome, Cytoplasmic vesicle, secretory vesicle, Cell projection, axon, Cell membrane, Presynapse, Postsynapse. Predicted band size: 25 kDa.
Rab proteins belong to the Ras small GTPase superfamily. The GTP-binding Rab3 subfamily has 4 known isoforms: 3A, 3B, 3C, and 3D. These proteins are expressed in highest concentrations in brain and endocrine tissues and involved in regulated exocytosis of hormones and neurotransmitters. Specifically, Rab 3s are thought to play a key role in presynaptic vesicle trafficking and priming.
Specifications
RAB3A | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P20336, P63011, P63012 | |
RAB3A | |
Synthetic peptide within human rab3a aa 121-170/220. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
PSH01-08 | |
Unconjugated | |
RAB3A | |
C18A3.6; RAB3; Rab3a; RAB3A, member RAS oncogene family; RAS-associated protein RAB3A; ras-related protein Rab-3A; ras-related protein Rab-3A-like protein; Ras-related small GTP binding protein 3A | |
Rabbit | |
Protein A | |
RUO | |
19339, 25531, 5864 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.