Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB3B Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297202PE
Description
RAB3B Polyclonal antibody specifically detects RAB3B in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
RAB3B | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
brain antigen RAB3B, RAB3B, member RAS oncogene family, ras-related protein Rab-3B | |
Produced in rabbits immunized with E. coli-derived Human RAB3B fragment. | |
0.1 mL | |
Signal Transduction | |
5865 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction