Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB3B Monoclonal Antibody (1543CT354.60.92)
GREENER_CHOICE

Catalog No. PIMA524738
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524738 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524738 Supplier Invitrogen™ Supplier No. MA524738
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RAB3B Monoclonal Antibody for Western Blot, IHC (P), Flow

Protein transport. Probably involved in vesicular traffic. Target protein may play a role in secretory granule function and exocytosis in pancreatic acinar cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB3B
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1543CT354.60.92
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Rab3b
Gene Accession No. P20337, Q63941, Q9CZT8
Gene Alias 2610528C18Rik; brain antigen RAB3B; Rab3b; RAB3B, member RAS oncogene family; ras-related protein Rab-3B
Gene Symbols Rab3b
Host Species Mouse
Immunogen human RAB3B recombinant protein.
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5865, 69908, 81755
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.