Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB3d Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587863
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587863 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587863 Supplier Invitrogen™ Supplier No. PA587863
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MASAGDTQAG PRDAADQNFD YMFKLLLIGN SSVGKTSFLF RYADDSFTPA FVSTVGIDFK VKTVYRHDKR IKLQIWDTAG QERYRTITTA YYRGAMGFLL MYDIANQESF AAVQDWATQI KTYSWDNAQV ILVGNKCDLE DERVVPAEDG RRLADDLGFE FFEASAKENI NVKQVFERLV DVICEKMNES LEPSSSSGSN GKGPAVGDAP APQPSSCSC; Positive Samples: HepG2, U-87MG, MCF7, SKOV3, Mouse lung, Mouse pancreas; Cellular Location: Cell membrane, Cytoplasmic side, Lipid-anchor.

Rab proteins are low-molecular-weight GTP-binding proteins that form the largest branch of the Ras superfamily of GTPases. Located on the cytoplasmic face of organelles and vesicles, rab proteins are involved in intracellular membrane fusion reactions. Three membrane proteins, synaptosomal associated protein of 25 kDa (SNAP-25), synaptobrevin, and syntaxin, form the core of a ubiquitous membrane fusion machine that interacts with the soluble proteins N-ethylmaleimide-sensitive factor (NSF) and a-SNAP. Rab proteins, in coordination with the core fusion machinery and Munc-18, help to mediate vesicle docking and fusion. There exist over 40 rab proteins that have been described in mammals. Rab 3D has been shown to be present, in part, in the tongue, pancreas, and liver.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB3d
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.94 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Rab3d
Gene Accession No. O95716, P35276
Gene Alias C130057E11Rik; D2-2; glioblastoma overexpressed; GOV; GTP-binding protein Rab-3D; RAB16; Rab3d; Rab3D upregulated with myeloid differentiation; RAB3D, member RAS oncogene family; RAD3D; Ras-related protein Rab-3D
Gene Symbols Rab3d
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-219 of human RAB3D (NP_0042741).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19340, 9545
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.