Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB3GAP1 Mouse anti-Human, Mouse, Rat, Clone: 4A3A1, Proteintech
SDP

Catalog No. 501737105 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7105 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7105 Supplier Proteintech Group Inc Supplier No. 666881IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RAB3GAP1, is a 981 amino acid protein that belongs to the Rab3-GAP catalytic subunit family. Rab 3 GAP p130 converts active RAB3-GTP to the inactive form RAB3-GDP, and is required for normal eye and brain development. Defects in Rab 3 GAP p130 are the cause of Warburg micro syndrome 1 (WARBM1). WARBM1 is a severe autosomal recessive disorder characterized by developmental abnormalities of the eye and central nervous system and by microgenitalia. The Rab 3 GAP p130 protein may participate in neurodevelopmental processes such as proliferation, migration and differentiation before synapse formation, and non-synaptic vesicular release of neurotransmitters.

Specifications

Antigen RAB3GAP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 4A3A1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene RAB3GAP1
Gene Accession No. P69735, Q15042, Q80UJ7
Gene Alias DKFZp434A012, KIAA0066, P130, Rab3 GAP, Rab3 GAP p130, RAB3GAP, RAB3GAP1, RAB3GAP130, WARBM1
Gene Symbols Rab3gap1
Host Species Mouse
Immunogen RAB3GAP1 Fusion Protein Ag17710
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 226407, 22930, 304759
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.