Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB3GAP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556952
Description
Immunogen sequence: PISELHLATT WPHLTEGIIV DNDVYSDLDP IQAPHWSVRV RKAENPQCLL GDFVTEFFKI CRRKESTDEI LGRSAFEEEG KETADITHAL SK Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.
RAB3GAP1, is a 981 amino acid protein that belongs to the Rab3-GAP catalytic subunit family. Rab 3 GAP p130 converts active RAB3-GTP to the inactive form RAB3-GDP, and is required for normal eye and brain development. Defects in Rab 3 GAP p130 are the cause of Warburg micro syndrome 1 (WARBM1). WARBM1 is a severe autosomal recessive disorder characterized by developmental abnormalities of the eye and central nervous system and by microgenitalia. The Rab 3 GAP p130 protein may participate in neurodevelopmental processes such as proliferation, migration and differentiation before synapse formation, and non-synaptic vesicular release of neurotransmitters.
Specifications
RAB3GAP1 | |
Polyclonal | |
Unconjugated | |
RAB3GAP1 | |
1700003B17Rik; 4732493F09Rik; AL117896; KIAA0066; P130; Rab 3 GAP p130; Rab3 GAP p130; RAB3 GTPase activating protein catalytic subunit 1; RAB3 GTPase activating protein subunit 1; RAB3 GTPase activating protein subunit 1 (catalytic); RAB3 GTPase-activating protein 130 kDa subunit; Rab3 GTPase-activating protein catalytic subunit; RAB3GAP; Rab3-GAP; Rab3-GAP p130; Rab3gap1; RAB3GAP130; RGD1306487; WARBM1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22930 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.8 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15042 | |
RAB3GAP1 | |
Recombinant protein corresponding to Human RAB3GAP1. Recombinant protein control fragment (Product #RP-102619). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction