Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB3GAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120651
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA5120651 100 μL
1 options
This item is not returnable. View return policy

Catalog No. PIPA5120651

Supplier: Invitrogen™ PA5120651

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HeLa, HepG2, Jurkat, Mouse lung, Mouse brain, Rat brain. Immunogen sequence: GLASGPPDLR CCLLHQKLQM LNCCIERKKA RDEGKKTSAS DVTNIYPGDA GKAGDQLVPD NLKETDKEKG EVGKSWDSWS DSEEEFFECL SDTEELKGNG QESGKKGGPK EMANLRPEGR LYQHGKLTLL HNGEPLYIPV TQEPAPMTED LLEEQSEVLA KLGTSAEGAH LRARMQSACL LSDMESFKAA NPGCSLEDFV RWYSPRDYIE EEVIDEKGNV VLKGELSARM KIPSNMWVEA WETAKPIPAR RQRRLFDDTR EAEKVLHYLA IQKPADLARH L.

RAB3GAP1, is a 981 amino acid protein that belongs to the Rab3-GAP catalytic subunit family. Rab 3 GAP p130 converts active RAB3-GTP to the inactive form RAB3-GDP, and is required for normal eye and brain development. Defects in Rab 3 GAP p130 are the cause of Warburg micro syndrome 1 (WARBM1). WARBM1 is a severe autosomal recessive disorder characterized by developmental abnormalities of the eye and central nervous system and by microgenitalia. The Rab 3 GAP p130 protein may participate in neurodevelopmental processes such as proliferation, migration and differentiation before synapse formation, and non-synaptic vesicular release of neurotransmitters.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB3GAP1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RAB3GAP1
Gene Accession No. P69735, Q15042, Q80UJ7
Gene Alias 1700003B17Rik; 4732493F09Rik; AL117896; KIAA0066; P130; Rab 3 GAP p130; Rab3 GAP p130; RAB3 GTPase activating protein catalytic subunit 1; RAB3 GTPase activating protein subunit 1; RAB3 GTPase activating protein subunit 1 (catalytic); RAB3 GTPase-activating protein 130 kDa subunit; Rab3 GTPase-activating protein catalytic subunit; RAB3GAP; Rab3-GAP; Rab3-GAP p130; Rab3gap1; RAB3GAP130; RGD1306487; WARBM1
Gene Symbols RAB3GAP1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 500-780 of human RAB3GAP1 (NP_001165906.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 226407, 22930, 304759
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.