Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB3GAP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA539385
Description
Rab 3 GAP p130, also known as Rab3 GTPase-activating protein catalytic subunit, is a 981 amino acid protein that belongs to the Rab3-GAP catalytic subunit family. Rab 3 GAP p130 converts active RAB3-GTP to the inactive form RAB3-GDP, and is required for normal eye and brain development. Defects in Rab 3 GAP p130 are the cause of Warburg micro syndrome 1 (WARBM1). WARBM1 is a severe autosomal recessive disorder characterized by developmental abnormalities of the eye and central nervous system and by microgenitalia. The Rab 3 GAP p130 protein may participate in neurodevelopmental processes such as proliferation, migration and differentiation before synapse formation, and non-synaptic vesicular release of neurotransmitters.
Specifications
RAB3GAP1 | |
Polyclonal | |
Unconjugated | |
RAB3GAP1 | |
Rab3 GTPase-activating protein catalytic subunit, RAB3 GTPase-activating protein 130 kDa subunit, Rab3-GAP p130, Rab3-GAP, KIAA0066, RAB3GAP, Rab 3 GAP p130, Rab3 GAP p130 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22930 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q15042 | |
RAB3GAP1 | |
A synthetic peptide derived from the internal region of human RAB3GAP1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction