Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB3GAP2 Mouse anti-Human, Clone: 3A4A1, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | RAB3GAP2 |
---|---|
Clone | 3A4A1 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-7066
![]() ![]() |
Proteintech Group Inc
666481IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The protein encoded by this gene belongs to the RAB3 protein family, members of which are involved in regulated exocytosis of neurotransmitters and hormones. This protein forms the Rab3 GTPase-activating complex with RAB3GAP1, where it constitutes the regulatory subunit, whereas the latter functions as the catalytic subunit. This gene has the highest level of expression in the brain, consistent with it having a key role in neurodevelopment. Mutations in this gene are associated with Martsolf syndrome.Specifications
RAB3GAP2 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
DKFZp434D245, FLJ14579, KIAA0839, p150, Rab3 GAP p150, Rab3 GAP regulatory subunit, RAB3 GAP150, RAB3GAP150, RAB3GAP2, RGAP iso | |
RAB3GAP2 | |
IgG2a | |
Protein A | |
Antibody |
3A4A1 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q9H2M9 | |
25782 | |
RAB3GAP2 Fusion Protein Ag20254 | |
Primary | |
-20°C | |
RAB3GAP2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title