Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB3GAP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101804
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101804 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101804 Supplier Invitrogen™ Supplier No. PA5101804
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RAB3GAP2.

The protein encoded by this gene belongs to the RAB3 protein family, members of which are involved in regulated exocytosis of neurotransmitters and hormones. This protein forms the Rab3 GTPase-activating complex with RAB3GAP1, where it constitutes the regulatory subunit, whereas the latter functions as the catalytic subunit. This gene has the highest level of expression in the brain, consistent with it having a key role in neurodevelopment. Mutations in this gene are associated with Martsolf syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB3GAP2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RAB3GAP2
Gene Accession No. Q5U1Z0, Q8BMG7, Q9H2M9
Gene Alias 1110059F07Rik; 2010002H18Rik; 5830469C09; AI851069; AW743433; KIAA0839; mKIAA0839; p150; RAB3 GTPase activating non-catalytic protein subunit 2; RAB3 GTPase activating protein subunit 2; RAB3 GTPase activating protein subunit 2 (non-catalytic); Rab3 GTPase-activating protein 150 kDa subunit; Rab3 GTPase-activating protein non-catalytic subunit; rab3-GAP p150; Rab3-GAP regulatory subunit; RAB3GAP150; RAB3-GAP150; RAB3GAP2; RGAP-iso; RGD1311518; SPG69; WARBM2
Gene Symbols RAB3GAP2
Host Species Rabbit
Immunogen A synthesized peptide derived from human RAB3GAP2(Accession Q9H2M9), corresponding to amino acid residues A268-A318.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25782, 289350, 98732
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.