Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB3GAP2 Polyclonal Antibody
RAB3GAP2 Polyclonal Antibody
Supplier: Thermo Scientific PA555574
Description
RAB3GAP2 Polyclonal Antibody for IHC

Specifications
RAB3GAP2 | |
Polyclonal | |
Unconjugated | |
RAB3GAP2 | |
1110059F07Rik; 2010002H18Rik; 5830469C09; AI851069; AW743433; KIAA0839; mKIAA0839; p150; RAB3 GTPase activating non-catalytic protein subunit 2; RAB3 GTPase activating protein subunit 2; RAB3 GTPase activating protein subunit 2 (non-catalytic); Rab3 GTPase-activating protein 150 kDa subunit; Rab3 GTPase-activating protein non-catalytic subunit; rab3-GAP p150; Rab3-GAP regulatory subunit; RAB3GAP150; RAB3-GAP150; RAB3GAP2; RGAP-iso; RGD1311518; SPG69; WARBM2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
25782 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H2M9 | |
RAB3GAP2 | |
Recombinant protein corresponding to Human RAB3GAP2. Recombinant protein control fragment (Product #RP-94142). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction