Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB3IL1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | RAB3IL1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
RAB3IL1 Polyclonal specifically detects RAB3IL1 in Human samples. It is validated for Western Blot.Specifications
| RAB3IL1 | |
| Polyclonal | |
| Rabbit | |
| Q8TBN0 | |
| 5866 | |
| Synthetic peptides corresponding to RAB3IL1(RAB3A interacting protein (rabin3)-like 1) The peptide sequence was selected from the middle region of RAB3IL1. Peptide sequence PTVKVAEVDCSSTNTCALSGLTRTCRHRIRLGDSKSHYYISPSSRARITA. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| GRAB, guanine nucleotide exchange factor for Rab3A, guanine nucleotide exchange factor for Rab-3A, RAB3A interacting protein (rabin3)-like 1, Rab-3A-interacting-like protein 1, Rab3A-interacting-like protein 1, Rabin3-like 1 | |
| RAB3IL1 | |
| IgG | |
| 43 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title