Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB3IL1 Antibody (OTI1B5), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273774V
Description
RAB3IL1 Monoclonal specifically detects RAB3IL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.Specifications
RAB3IL1 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
RAB3IL1 | |
Full length recombinant protein of human RAB3IL1 (NP_037533) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Flow Cytometry | |
OTI1B5 | |
Western Blot, Flow Cytometry | |
GRAB, guanine nucleotide exchange factor for Rab3A, guanine nucleotide exchange factor for Rab-3A, RAB3A interacting protein (rabin3)-like 1, Rab-3A-interacting-like protein 1, Rab3A-interacting-like protein 1, Rabin3-like 1 | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
5866.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction