Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB3IL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558901
Description
Immunogen sequence: MDSSEEHAGC PARGTCPVFL AMSAGTVRYA PSGLCPVLEG NLREEPWGTD SPPQPDQGLP PPLAAVPV Highest antigen sequence identity to the following orthologs: Mouse - 32%, Rat - 32%.
Guanine nucleotide exchange factor (GEF) for Rab3A, a GTPase that regulates synaptic vesicle exocytosis.
Specifications
RAB3IL1 | |
Polyclonal | |
Unconjugated | |
Rab3il1 | |
1200014K04Rik; AI115013; C76746; Grab; guanine nucleotide exchange factor for Rab3A; Guanine nucleotide exchange factor for Rab-3A; RAB3A interacting protein (rabin3)-like 1; RAB3A interacting protein like 1; Rab3ail1; rab-3A-interacting-like protein 1; rab3A-interacting-like protein 1; RAB3IL1; Rabin3-like 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5866 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TBN0 | |
Rab3il1 | |
Recombinant protein corresponding to Human RAB3IL1. Recombinant protein control fragment (Product #RP-97443). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction