Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB3IP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | RAB3IP |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Regulatory Status | RUO |
Description
RAB3IP Polyclonal specifically detects RAB3IP in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
RAB3IP | |
Unconjugated | |
RUO | |
FLJ14660, FLJ22548, MGC71495, RAB3A interacting protein (rabin3), rab-3A-interacting protein, Rab3A-interacting protein, rabin-3, RABIN3, SSX2 interacting protein, SSX2-interacting protein | |
RAB3IP | |
IgG |
Polyclonal | |
Rabbit | |
Q96QF0 | |
117177 | |
Synthetic peptides corresponding to RAB3IP(RAB3A interacting protein (rabin3)) The peptide sequence was selected from the N terminal of RAB3IP. Peptide sequence SPDLLGVYESGTQEQTTSPSVIYRPHPSALSSVPIQANALDVSELPTQPV. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title