Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB3IP Antibody (OTI5F2), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273775AF700
Description
RAB3IP Monoclonal specifically detects RAB3IP in Human, Mouse, Rat, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry.Specifications
RAB3IP | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
RAB3IP | |
Full length recombinant protein of human RAB3IP (NP_071901) produced in HEK293T cells. | |
0.1 mL | |
117177.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI5F2 | |
Western Blot, Immunohistochemistry | |
FLJ14660, FLJ22548, MGC71495, RAB3A interacting protein (rabin3), rab-3A-interacting protein, Rab3A-interacting protein, rabin-3, RABIN3, SSX2 interacting protein, SSX2-interacting protein | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Zebrafish | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction