Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB3IP Monoclonal Antibody (OTI5H1), TrueMAB™, OriGene
SDP

Catalog No. 501682505
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-2505 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-2505 Supplier OriGene Supplier No. TA808986
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The specific function of this protein remains unknown.

Specifications

Antigen RAB3IP
Applications Western Blot
Classification Monoclonal
Clone OTI5H1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene RAB3IP
Gene Accession No. Q96QF0
Gene Alias RABIN3, RABIN8
Gene Symbols RAB3IP
Host Species Mouse
Immunogen Full length human recombit protein of human RAB3IP produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117177
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.