Learn More
RAB3IP Mouse anti-Human, Clone: OTI4A7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF808962

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The specific function of this protein remains unknown.Specifications
RAB3IP | |
Monoclonal | |
Unconjugated | |
RAB3IP | |
RABIN3, RABIN8 | |
Mouse | |
Affinity Chromatography | |
RUO | |
117177 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI4A7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q96QF0 | |
RAB3IP | |
Full length human recombit protein of human RAB3IP produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.