Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB3IP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588659
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588659 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588659 Supplier Invitrogen™ Supplier No. PA588659
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MANDPLEGFH EVNLASPTSP DLLGVYESGT QEQTTSPSVI YRPHPSALSS VPIQANALDV SELPTQPVYS SPRRLNCAEI SSISFHVTDP APCSTSGVTA GLTKLTTRKD NYNAEREFLQ GATITEACDG SDDIFGLSTD SLSRLRSPSV; Positive Samples: 293T; Cellular Location: Cell projection, Cytoplasm, Nucleus, cytoskeleton, lamellipodium.

Guanine nucleotide exchange factor (GEF) which may activate RAB8A and RAB8B. Promotes the exchange of GDP to GTP, converting inactive GDP-bound Rab proteins into their active GTP-bound form. Mediates the release of GDP from RAB8A and RAB8B but not from RAB3A or RAB5. Modulates actin organization and promotes polarized transport of RAB8A-specific vesicles to the cell surface. Together with RAB11A, RAB8A, the exocyst complex, PARD3, PRKCI, ANXA2, CDC42 and DNMBP promotes transcytosis of PODXL to the apical membrane initiation sites (AMIS), apical surface formation and lumenogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB3IP
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RAB3IP
Gene Accession No. Q62739, Q68EF0, Q96QF0
Gene Alias B230311A06; gene trap PAT 12; Gtpat12; MGC137223 protein; RAB3A interacting protein; RAB3A interacting protein (rabin3); rab-3A-interacting protein; RAB3A-interacting protein; RAB3IP; rab3ip {ECO:0000250; rabin 3; Rabin3; rabin-3; RABIN8; SSX2 interacting protein; SSX2-interacting protein; UniProtKB:Q96QF0}
Gene Symbols RAB3IP
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human RAB3IP (NP_0719012).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117177, 216363, 29885
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.