Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rab4 Monoclonal Antibody (4E11)
GREENER_CHOICE

Catalog No. PIMA517161
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517161 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517161 Supplier Invitrogen™ Supplier No. MA517161
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-17161 targets RAB4A in FACS, IHC, indirect ELISA, and WB applications and shows reactivity with Human, Mouse, and Non-human primate samples. The MA5-17161 immunogen is purified recombinant fragment of human RAB4A expressed in E. Coli. MA5-17161 detects RAB4A which has a predicted molecular weight of approximately 23.8kDa.

Protein transport. Probably involved in vesicular traffic.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rab4
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 4E11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene Rab4a
Gene Accession No. P20338, P56371
Gene Alias AI848268; HRES1; HRES-1; HRES-1/RAB4; HTLV-1 related endogenous sequence; RAB4; Rab4a; RAB4A, member RAS oncogene family; ras-related protein Rab-4A; Ras-related small GTP binding protein 4
Gene Symbols Rab4a
Host Species Mouse
Immunogen Purified recombinant fragment of human RAB4A expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19341, 5867
Target Species Human, Mouse, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.