Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987087
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7087 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7087 Supplier Bioss Supplier No. BS6157R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Protein transport. Probably involved in vesicular traffic (By similarity).

Specifications

Antigen RAB4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Rab4a
Gene Accession No. P20338
Gene Alias AI848268; HRES1; HRES-1; HRES-1/RAB4; HTLV-1 related endogenous sequence; Rab4; Rab4a; RAB4A, member RAS oncogene family; Ras-related protein Rab-4A; Ras-related small GTP binding protein 4
Gene Symbols Rab4a
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human RAB4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5867
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.