Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB40A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104078
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104078 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104078 Supplier Invitrogen™ Supplier No. PA5104078
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RAB40A.

This gene encodes a member of the Rab40 subfamily of Rab small GTP-binding proteins that contains a C-terminal suppressors of cytokine signaling box.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB40A
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RAB40A
Gene Accession No. Q8WXH6
Gene Alias protein Rar-2; RAB40A; RAB40A, member RAS oncogene family; RAR2; RAR2A; Ras-related protein Rab-40A; SOCS box containing protein RAR2A; SOCS box-containing protein RAR2A
Gene Symbols RAB40A
Host Species Rabbit
Immunogen A synthesized peptide derived from human RAB40A(Accession Q8WXH6), corresponding to amino acid residues T244-S277.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 142684
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.