Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB40A Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988501
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8501 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8501 Supplier Bioss Supplier No. BS8366R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a member of the Rab40 subfamily of Rab small GTP-binding proteins that contains a C-terminal suppressors of cytokine signaling box.

Specifications

Antigen RAB40A
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RAB40A
Gene Accession No. Q8WXH6
Gene Alias Protein Rar-2; RAB40A; RAB40A, member RAS oncogene family; RAR2; RAR2A; Ras-related protein Rab-40A; SOCS box containing protein RAR2A; SOCS box-containing protein RAR2A
Gene Symbols RAB40A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human RAB40A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 142684
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.