Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB40B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536509
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536509 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536509 Supplier Invitrogen™ Supplier No. PA536509
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 34 kDa. Purity is >95% by SDS-PAGE.

Rab 40B is a 278 amino acid protein that belongs to the small GTPase superfamily and the Rab family. Rab 40B contains a SOCS box domain that mediates interactions with the Elongin BC complex, an adapter module in different E3 ubiquitin ligase complexes. Rab 40B shares similarity with a yeast protein, suggesting a role in regulation of secretory vesicles. The Rab 40B gene is conserved in chimpanzee, cow, mouse, rat, chicken and zebrafish, and maps to human chromosome 17q25.3. It has been suggested that there is a major gene locus on chromosome 17q25.3 for autosomal dominant moyamoya disease (MMD), which is an idiopathic steno-occlusive cerebrovascular disease that represents an important cause of stroke.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB40B
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Rab40b
Gene Accession No. Q12829, Q8VHP8
Gene Alias GTP-binding protein homologous to Saccharomyces cerevisiae SEC4; protein Rar; RAB40B; Rab40b, member RAS oncogene family; rar; RAR SOCS box containing protein; ras-related protein Rab-40B; sec4; SEC4L; SOCS box-containing protein RAR
Gene Symbols Rab40b
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 231-278 of Human Rab 40B.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10966, 217371, 303754
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.