Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB40B Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501728004 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8004 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8004 Supplier Proteintech Group Inc Supplier No. 124711AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Rab 40B is a 278 amino acid protein that belongs to the small GTPase superfamily and the Rab family. Rab 40B contains a SOCS box domain that mediates interactions with the Elongin BC complex, an adapter module in different E3 ubiquitin ligase complexes. Rab 40B shares similarity with a yeast protein, suggesting a role in regulation of secretory vesicles. The Rab 40B gene is conserved in chimpanzee, cow, mouse, rat, chicken and zebrafish, and maps to human chromosome 17q25.3. It has been suggested that there is a major gene locus on chromosome 17q25.3 for autosomal domit moyamoya disease (MMD), which is an idiopathic steno-occlusive cerebrovascular disease that represents an important cause of stroke.

Specifications

Antigen RAB40B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene RAB40B
Gene Accession No. Q12829
Gene Alias Protein Rar, RAB40B, RAR, Ras related protein Rab 40B, SEC4L
Gene Symbols RAB40B
Host Species Rabbit
Immunogen RAB40B Fusion Protein Ag3155
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10966
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.