Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RAB40C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104080
Description
Antibody detects endogenous levels of total RAB40C.
RAB40C is a probable substrate-recognition component of a SCF-like ECS (Elongin-Cullin-SOCS-box protein) E3 ubiquitin ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins.
Specifications
RAB40C | |
Polyclonal | |
Unconjugated | |
RAB40C | |
cb453; rab40c; Rab40c, member RAS oncogene family; RAR (RAS like GTPASE) like; RAR like; RAR3; RARL; rar-like protein; RASL8C; RAS-like GTPase; ras-like protein family member 8C; RAS-like, family 8, member C; ras-related protein Rab-40C; SOCS box containing protein RAR3; SOCS box-containing protein RAR3; Unknown (protein for MGC:136966); wu:fk50d06; zgc:136966 | |
Rabbit | |
Affinity chromatography | |
RUO | |
224624, 359728, 57799 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8VHQ4, Q96S21 | |
RAB40C | |
A synthesized peptide derived from human RAB40C(Accession Q96S21), corresponding to amino acid residues G246-S281. | |
100 μL | |
Primary | |
Human, Mouse, Rat, Monkey | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction