Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAB40C Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988502
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8502 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8502 Supplier Bioss Supplier No. BS8367R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RAB40C is a probable substrate-recognition component of a SCF-like ECS (Elongin-Cullin-SOCS-box protein) E3 ubiquitin ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins.

Specifications

Antigen RAB40C
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RAB40C
Gene Accession No. Q96S21
Gene Alias BOS_22518; cb453; Rab40c; RAB40C, member RAS oncogene family; RAR (RAS like GTPASE) like; RAR like; RAR3; RARL; Rar-like protein; RASL8C; RAS-like GTPase; Ras-like protein family member 8C; RAS-like, family 8, member C; Ras-related protein Rab-40C; SOCS box containing protein RAR3; SOCS box-containing protein RAR3; Unknown (protein for MGC:136966); wu:fk50d06; zgc:136966
Gene Symbols RAB40C
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human RAB40C.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57799
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.