Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB41 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551592
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551592 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551592 Supplier Invitrogen™ Supplier No. PA551592
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GKTSIISRFM YNSFGCACQA TVGIDFLSKT MYLEDQIVQL QLWDTAGQER FHSLIPSYIR DSTIAVVVYD ITNINSFKET DKWVEHVRAE RGDDVVIMLL GNKIDLDNKR QVTAEQGEEK SRNLNVMFIE TSAKTGYNVK KVI Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 73%.

Rab 41 (Ras-related protein Rab-41) is a 222 amino acid protein that belongs to the small GTPase superfamily and the Rab family.Members of the Rab familyfunction asregulators ofmembrane trafficking. They cycle between inactive GDP-bound and activated GTP-bound states, which is controlled by GTP hydrolysis-activating proteins (GAPs). Rab 41 can be activated by the GAP protein RN-Tre. Localizing to the Golgi complex, Rab 41 is involved in protein transport and participates in GTP binding. Existing as two alternatively spliced isoforms, the gene that encodes the Rab 41 protein maps to human chromosome Xq13.1. Chromosome X consists of about 153 million base pairs and nearly 1,000 genes. Color blindness, hemophilia, and Duchenne muscular dystrophy are well known X chromosome-linked conditions which affect males more frequently as males carry a single X chromosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB41
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RAB41
Gene Accession No. Q5JT25
Gene Alias RAB41; RAB41, member RAS homolog family; RAB41, member RAS oncogene family; ras-related protein Rab-41
Gene Symbols RAB41
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RAB41. Recombinant protein control fragment (Product #RP-92007).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 347517
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.