Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB42 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565606
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565606 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565606 Supplier Invitrogen™ Supplier No. PA565606
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GVRLIHKTQIP RSPSRKQHSG PCQC Highest antigen sequence identity to the following orthologs - mouse 68%, rat 72%.

RAB42 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include membrane; GTP binding; GTPase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB42
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RAB42
Gene Accession No. Q8N4Z0
Gene Alias 9530096D07Rik; LOW QUALITY PROTEIN: ras-related protein Rab-42; putative Ras-related protein Rab-42; RAB42; RAB42, member RAS homolog family; RAB42, member RAS oncogene family; RAB42, member RAS oncogene family, pseudogene; Rab42-ps; Rab43; Ras-related protein Rab-42
Gene Symbols RAB42
Host Species Rabbit
Immunogen Recombinant Human RAB42. Recombinant protein control fragment (Product #RP-106270).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115273
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.