Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB44 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145212
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145212 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145212 Supplier Invitrogen™ Supplier No. PA5145212
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GQERYHSMTR QLLRKADGVV LMYDITSQES FAHVRYWLDC LQDAGSDGVV ILLLGNKMDC EEERQVSVEA GQQLAQ Antigen sequence identity: Mouse 83%; Rat 82%.

RAB44 is a member of the RAS oncogene family of small GTPases that plays a role in intracellular vesicle trafficking and membrane fusion events. It is expressed in various tissues, including the brain, liver, and testis, and has been implicated in the regulation of autophagy and lysosomal function. However, its precise biological function and molecular mechanisms of action remain poorly understood.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB44
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RAB44
Gene Accession No. Q7Z6P3
Gene Symbols RAB44
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 891-966.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 401258
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.