Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAB4A Rabbit anti-Human, Unconjugated, Clone: CCI-18, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB20645
Description
RAB4A Monoclonal antibody specifically detects RAB4A in Human samples. It is validated for ICC, WB-Ce, IF, Flow Cyt.Specifications
RAB4A | |
Monoclonal | |
Unconjugated | |
RAB4A | |
Rabbit | |
100 μg | |
5867 | |
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
IgG |
Immunocytochemistry, Western Blot, Immunofluorescence, Flow Cytometry | |
CCI-18 | |
RAB4A, member RAS oncogene family | |
HRES-1/RAB4, RAB4 | |
A synthetic peptide corresponding to human RAB4A | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction