Learn More
Invitrogen™ RAB4B Polyclonal Antibody

Description
Predicted to react with zebrafish, mouse and rat based on sequence homology.
Specifications
Specifications
| Antigen | RAB4B |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | RAB4B |
| Gene Accession No. | P61018 |
| Gene Alias | 1500031G17Rik; PP1596; Rab4; RAB4B; RAB4B, member RAS oncogene family; ras-related GTP-binding protein 4b; ras-related protein Rab-4B; small GTP binding protein RAB4B |
| Gene Symbols | RAB4B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.