Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB4B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112680
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112680 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112680 Supplier Invitrogen™ Supplier No. PA5112680
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RAB4B Polyclonal Antibody for IHC (P), ELISA

Protein transport. Probably involved in vesicular traffic.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB4B
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene RAB4B
Gene Accession No. P61018, Q91ZR1
Gene Alias 1500031G17Rik; PP1596; Rab4; RAB4B; RAB4B, member RAS oncogene family; ras-related GTP-binding protein 4b; ras-related protein Rab-4B; small GTP binding protein RAB4B
Gene Symbols RAB4B
Host Species Rabbit
Immunogen Recombinant Human Ras-related protein Rab-4B protein (2-213AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19342, 53916
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.