Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAB5 Recombinant Rabbit Monoclonal Antibody (0K5J1)
GREENER_CHOICE

Catalog No. PIMA535161
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535161 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535161 Supplier Invitrogen™ Supplier No. MA535161
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: KELQRQASPN IVIALSGNKA DLANKRAVDF QEAQSYADDN SLLFMETSAK TSMNVNEIFM AIAKKLPKNE PQNPGANSAR GRGVDLTEPT QPTRNQCCSN.

RAB5 are among the small GTPases Rab are key regulators of intracellular membrane trafficking, from the formation of transport vesicles to their fusion with membranes. Rabs cycle between an inactive GDP-bound form and an active GTP-bound form that is able to recruit to membranes different sets of downstream effectors directly responsible for vesicle formation, movement, tethering and fusion. RAB5A is required for the fusion of plasma membranes and early endosomes. Contributes to the regulation of filopodia extension. Required for the exosomal release of SDCBP, CD63, PDCD6IP and syndecan. Regulates maturation of apoptotic cell-containing phagosomes, probably downstream of DYN2 and PIK3C3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB5
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 0K5J1
Concentration 0.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RAB5A
Gene Accession No. M0RC99, P20339, Q9CQD1
Gene Alias 2410015H04Rik; AI663973; AU021172; nnyRab5a; Rab 5; Rab 5A; RAB5; RAB5A; RAB5A, member RAS oncogene family; Ras related protein; RAS-associated protein RAB5A; ras-related protein Rab-5A; Small GTP-binding protein rab5
Gene Symbols RAB5A
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 116-215 of human RAB5AA (P20339).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 271457, 5868, 64633
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.